Learn More
Biomatik Corporation Human Nerve Growth Factor (NGF), Active Protein
Supplier:  Biomatik Corporation RPU54733
Encompass_Preferred
Human Nerve Growth Factor (NGF), Active Protein is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Nerve Growth Factor (NGF). Target Synonyms: NGFB; Beta-NGF; HSAN5; NGF-B; Beta-Nerve Growth Factor. Accession Number: P01138. Gene ID: 4803. Expression Region: Glu19~Arg239. Tag Info: N-terminal His Tag. Theoretical MW: 30/33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0250