Learn More
Biomatik Corporation Recombinant Human Glucose Transporter 1 (GLUT1)
Supplier:  Biomatik Corporation RPU54835
Encompass_Preferred
Recombinant Human Glucose Transporter 1 (GLUT1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Glucose Transporter 1 (GLUT1). Target Synonyms: SLC2A1; SLC2-A1; GLUT; Solute Carrier Family 2 Member 1,Facilitated Glucose Transporter; Glucose transporter type 1, erythrocyte/brain; HepG2 glucose transporter. Accession Number: P11166. Gene ID: 6513. Expression Region: Met251~Glu329. Tag Info: Two N-terminal Tags, His-tag and SUMO-tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0256