Learn More
Biomatik Corporation Human Mdm2 p53 Binding Protein Homolog (MDM2), Active Protein
Supplier:  Biomatik Corporation RPU54979
Encompass_Preferred
Human Mdm2 p53 Binding Protein Homolog (MDM2), Active Protein is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Mdm2 p53 Binding Protein Homolog (MDM2). Target Synonyms: HDMX; hdm2; Mouse Double Minute 2,Human Homolog Of p53-Binding Protein; Oncoprotein Mdm2. Accession Number: Q00987. Gene ID: 4193. Expression Region: Met1~Pro218. Tag Info: N-terminal His Tag. Theoretical MW: 44kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0274