Learn More
Biomatik Corporation Native Human Fibrinogen (FG)
Supplier:  Biomatik Corporation RPU55143
Encompass_Preferred
Native Human Fibrinogen (FG) is a purified Natural Protein. Purity: >97% by SDS-PAGE. Host: Human. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Fibrinogen (FG). Target Synonyms: F1; FI; Fbg; Coagulation Factor I. Accession Number: P02675. Gene ID: 2244. Theoretical MW: 70/56/52kDa. Buffer: PBS, pH7.4, containing 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0276