Learn More
Biomatik Corporation Recombinant Human Bone Sialoprotein (BSP)
Supplier:  Biomatik Corporation RPU55268
Encompass_Preferred
Recombinant Human Bone Sialoprotein (BSP) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Bone Sialoprotein (BSP). Target Synonyms: IBSP; Bsp; BspII; SpII; Integrin-Binding Bone Sialoprotein; Cell-binding sialoprotein; Bone sialoprotein 2. Accession Number: P21815. Gene ID: 3381. Expression Region: Gln174~Gln317. Tag Info: Two N-terminal Tags, His-tag and SUMO-tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0285