Learn More
Biomatik Corporation Recombinant Rat Colony Stimulating Factor 1, Macrophage (MCSF)
Supplier:  Biomatik Corporation RPU55478
Encompass_Preferred
Recombinant Rat Colony Stimulating Factor 1, Macrophage (MCSF) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Colony Stimulating Factor 1, Macrophage (MCSF). Target Synonyms: CSF1; M-CSF; Lanimostim; Processed macrophage colony-stimulating factor 1. Accession Number: Q8JZQ0. Expression Region: Glu261~Leu427. Tag Info: Two N-terminal Tags, His-tag and SUMO-tag. Theoretical MW: 46kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0300