Learn More
Biomatik Corporation Human Vascular Endothelial Growth Factor A (VEGFA), Active Protein
Supplier:  Biomatik Corporation RPU55499
Encompass_Preferred
Human Vascular Endothelial Growth Factor A (VEGFA), Active Protein is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Vascular Endothelial Growth Factor A (VEGFA). Target Synonyms: VEGF; VEGF-A; VPF; Vascular Endothelial Cell Growth Factor A; Vascular Permeability Factor. Accession Number: P15692. Gene ID: 7422. Expression Region: Pro28~Arg147. Tag Info: N-terminal His Tag. Theoretical MW: 17/18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0304