Learn More
Biomatik Corporation Recombinant Mouse Corticotropin Releasing Factor (CRF)
Supplier:  Biomatik Corporation RPU41019
Encompass_Preferred
Recombinant Mouse Corticotropin Releasing Factor (CRF) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Corticotropin Releasing Factor (CRF). Target Synonyms: CRH; Corticotropin Releasing Hormone; Corticoliberin. Accession Number: Q8CIT0. Gene ID: 12918. Expression Region: Arg28~Arg160. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9914