Learn More
Biomatik Corporation Mouse Receptor Activator Of Nuclear Factor Kappa B Ligand (RANkL), Active Protein
Supplier:  Biomatik Corporation RPU55616
Encompass_Preferred
Mouse Receptor Activator Of Nuclear Factor Kappa B Ligand (RANkL), Active Protein is a purified Eukaryotic Protein. Purity: >90% by SDS-PAGE. Host: 293F Cell. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Receptor Activator Of Nuclear Factor Kappa B Ligand (RANkL). Target Synonyms: CD254; TNFSF11; ODF; OPGL; TRANCE; HRANKL2; SOdf; Tumor Necrosis Factor(ligand)superfamily Member 11; TNF-related activation-induced cytokine. Accession Number: O35235. Gene ID: 21943. Expression Region: Arg72~Asp316. Tag Info: N-terminal His Tag. Theoretical MW: 35kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0311