Learn More
Biomatik Corporation Human Complement Component 1, Q Subcomponent A (C1qA), Active Protein
Supplier:  Biomatik Corporation RPU55829
Encompass_Preferred
Human Complement Component 1, Q Subcomponent A (C1qA), Active Protein is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Complement Component 1, Q Subcomponent A (C1qA). Target Synonyms: Complement Component 1,q Subcomponent,Alpha Polypeptide. Accession Number: P02745. Gene ID: 712. Expression Region: Ala28~Ala245. Tag Info: N-terminal His Tag. Theoretical MW: 30kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0317