Learn More
Biomatik Corporation Recombinant Mouse Gasdermin D (GSDMD)
Supplier:  Biomatik Corporation RPU55896
Encompass_Preferred
Recombinant Mouse Gasdermin D (GSDMD) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Gasdermin D (GSDMD). Target Synonyms: GSDMDC1; DF5L; Gasdermin Domain Containing 1. Accession Number: Q9D8T2. Gene ID: 69146. Expression Region: Met1~Gln325. Tag Info: N-terminal His Tag. Theoretical MW: 40kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0319