Learn More
Biomatik Corporation Recombinant Maltose Binding Protein (MBP)
Supplier:  Biomatik Corporation RPU56080
Encompass_Preferred
Recombinant Maltose Binding Protein (MBP) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: Maltose Binding Protein (MBP). Accession Number: P0AEX9. Gene ID: 75204178. Expression Region: Lys27~Thr392. Tag Info: N-terminal His Tag. Theoretical MW: 51kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0325