Learn More
Biomatik Corporation Recombinant Human Erythroferrone (ERFE)
Supplier:  Biomatik Corporation RPU56180
Encompass_Preferred
Recombinant Human Erythroferrone (ERFE) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Erythroferrone (ERFE). Target Synonyms: FAM132B; C1QTNF15; CTRP15; Myonectin; Family With Sequence Similarity 132, Member B; Complement C1q tumor necrosis factor-related protein 15. Accession Number: Q4G0M1. Gene ID: 151176. Expression Region: Ser29~Val354. Tag Info: N-terminal His Tag. Theoretical MW: 41kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0329