Learn More
Biomatik Corporation Recombinant Human Cluster Of differentiation 299 (CD299)
Supplier:  Biomatik Corporation RPU41047
Encompass_Preferred
Recombinant Human Cluster Of differentiation 299 (CD299) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cluster Of differentiation 299 (CD299). Target Synonyms: CLEC4M; CD209L; DC-SIGN2; DC-SIGNR; L-SIGN; C-Type Lectin Domain Family 4, Member M; CD209 like 1; Dendritic cell-specific ICAM-3-grabbing non-integrin 2; Liver/lymph node-specific ICAM-3-grabbing non. Accession Number: Q9H2X3. Gene ID: 10332. Expression Region: Pro242~Glu399. Tag Info: N-terminal His Tag. Theoretical MW: 23kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9916