Learn More
Biomatik Corporation Recombinant Human Interferon Kappa (IFNk)
Supplier:  Biomatik Corporation RPU57196
Encompass_Preferred
Recombinant Human Interferon Kappa (IFNk) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interferon Kappa (IFNk). Target Synonyms: IFN-K. Accession Number: Q9P0W0. Gene ID: 56832. Expression Region: Leu28~Lys207. Tag Info: N-terminal His and GST Tag. Theoretical MW: 52kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0343