Learn More
Biomatik Corporation Recombinant Human Taste Receptor Type 1 Member 3 (TAS1R3)
Supplier:  Biomatik Corporation RPU59581
Encompass_Preferred
Recombinant Human Taste Receptor Type 1 Member 3 (TAS1R3) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Taste Receptor Type 1 Member 3 (TAS1R3). Target Synonyms: T1R3; Sweet taste receptor T1R3. Accession Number: Q7RTX0. Gene ID: 83756. Expression Region: Ala21~Leu570. Tag Info: N-terminal His Tag. Theoretical MW: 65kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0362