Learn More
Biomatik Corporation Recombinant Human Bone morphogenetic protein 4 (BMP4)
Supplier:  Biomatik Corporation RPC20011
Encompass_Preferred
Recombinant Human Bone morphogenetic protein 4 (BMP4) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: BMP4. Target Synonyms: Bone morphogenetic protein 2B; BMP-2B. Accession Number: P12644. Expression Region: 293~408aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 29.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0365