Learn More
Biomatik Corporation Recombinant Mouse Cyclophilin B (CYPB)
Supplier:  Biomatik Corporation RPU41096
Encompass_Preferred
Recombinant Mouse Cyclophilin B (CYPB) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Cyclophilin B (CYPB). Target Synonyms: PPIB; PPI-B; CYP-S1; CyP-B; SCYLP; PPIase B; Rotamase B; S-cyclophilin; Peptidylprolyl Isomerase B; Peptidyl-Prolyl Cis-Trans Isomerase B. Accession Number: P24369. Gene ID: 19035. Expression Region: Asn34~Glu216. Tag Info: N-terminal His Tag. Theoretical MW: 21kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9920