Learn More
Biomatik Corporation Recombinant Human Protein Wnt-10a (WNT10A)
Supplier:  Biomatik Corporation RPC20272
Encompass_Preferred
Recombinant Human Protein Wnt-10a (WNT10A) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: WNT10A. Target Synonyms: FLJ14301; OODD; Protein Wnt-10a; SSPS; STHAG4; Wingless type MMTV integration site family member 10A; WN10A_HUMAN; wnt10a. Accession Number: Q9GZT5. Expression Region: 36~417aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 44.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0395