Learn More
Biomatik Corporation Recombinant Heteroscodra maculata Delta-theraphotoxin-Hm1a
Supplier:  Biomatik Corporation RPC20465
Encompass_Preferred
Recombinant Heteroscodra maculata Delta-theraphotoxin-Hm1a is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Heteroscodra maculata (Togo starburst tarantula) (Togo starburst baboon spider). Target Name: Heteroscodra maculata Delta-theraphotoxin-Hm1a. Target Synonyms: Delta-theraphotoxin-Hm1a; Delta-TRTX-Hm1a; Heteroscodratoxin-1; HmTx1; Kappa-theraphotoxin-Hm1a; Kappa-TRTX-Hm1a. Accession Number: P60992. Expression Region: 1~35aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 24kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0411