Learn More
Biomatik Corporation Recombinant Human Elongation factor 1-alpha 1 (EEF1A1)
Supplier:  Biomatik Corporation RPC20579
Encompass_Preferred
Recombinant Human Elongation factor 1-alpha 1 (EEF1A1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: EEF1A1. Target Synonyms: Elongation factor Tu. Accession Number: P68104. Expression Region: 1~462aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 66.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0413