Learn More
Biomatik Corporation Recombinant Human Thioredoxin-interacting protein (TXNIP)
Supplier:  Biomatik Corporation RPC20839
Encompass_Preferred
Recombinant Human Thioredoxin-interacting protein (TXNIP) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: In Vitro E. coli Expression System. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: TXNIP. Target Synonyms: Thioredoxin-binding protein 2Vitamin D3 up-regulated protein 1. Accession Number: Q9H3M7. Expression Region: 1~391aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 57.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0428