Learn More
Biomatik Corporation Recombinant Human Inhibitory Subunit Of NF Kappa B Alpha (IkBa)
Supplier:  Biomatik Corporation RPU42013
Encompass_Preferred
Recombinant Human Inhibitory Subunit Of NF Kappa B Alpha (IkBa) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Inhibitory Subunit Of NF Kappa B Alpha (IkBa). Target Synonyms: NFKBIA; IKB-A; MAD-3; NFKBI; Nuclear Factor Of Kappa Light Polypeptide Gene Enhancer In B-Cells Inhibitor,Alpha; Major histocompatibility complex enhancer-binding protein MAD3. Accession Number: P25963. Gene ID: 4792. Expression Region: Gly74~Thr247. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9981