Learn More
Biomatik Corporation Recombinant Human Cytochrome P450 2D6 (CYP2D6)
Supplier:  Biomatik Corporation RPU41129
Encompass_Preferred
Recombinant Human Cytochrome P450 2D6 (CYP2D6) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cytochrome P450 2D6 (CYP2D6). Target Synonyms: CYP2D; P450C2D; CPD6; CYP2DL1; P450-DB1; Cytochrome P450 Family 2 Subfamily D Polypeptide 6; Cytochrome P450-DB1; Debrisoquine 4-hydroxylase. Accession Number: P10635. Gene ID: 1565. Expression Region: Leu236~Gln472. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9924