Learn More
Biomatik Corporation Recombinant Human Tar DNA Binding Protein 43kDa (TDP43)
Supplier:  Biomatik Corporation RPU43947
Encompass_Preferred
Recombinant Human Tar DNA Binding Protein 43kDa (TDP43) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Tar DNA Binding Protein 43kDa (TDP43). Target Synonyms: TARDBP; ALS10. Accession Number: Q13148. Gene ID: 23435. Expression Region: Ser104~Pro262. Tag Info: N-terminal His Tag. Theoretical MW: 21/19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0060