Learn More
Biomatik Corporation Recombinant Human Actinin Alpha 1 (ACTN1)
Supplier:  Biomatik Corporation RPU50422
Encompass_Preferred
Recombinant Human Actinin Alpha 1 (ACTN1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Actinin Alpha 1 (ACTN1). Target Synonyms: Alpha-actinin cytoskeletal isoform; F-actin cross-linking protein; Non-muscle alpha-actinin-1. Accession Number: P12814. Gene ID: 87. Expression Region: Ala28~Asn260. Tag Info: N-terminal His Tag. Theoretical MW: 30kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0101