Learn More
Biomatik Corporation Recombinant Mouse Sjogren Syndrome Antigen A1 (SSA1)
Supplier:  Biomatik Corporation RPU50542
Encompass_Preferred
Recombinant Mouse Sjogren Syndrome Antigen A1 (SSA1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Sjogren Syndrome Antigen A1 (SSA1). Target Synonyms: RO52; RNF81; RO52; TRIM21; 52kDa,Ribonucleoprotein Autoantigen SS-A/Ro; Tripartite Motif-Containing 21; Autoantigen Ro, 52kDa; RING finger protein 81. Accession Number: Q62191. Expression Region: Asp272~Met470. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0107