Learn More
Biomatik Corporation Recombinant Rat Glial Fibrillary Acidic Protein (GFAP)
Supplier:  Biomatik Corporation RPU50610
Encompass_Preferred
Recombinant Rat Glial Fibrillary Acidic Protein (GFAP) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Glial Fibrillary Acidic Protein (GFAP). Target Synonyms: Intermediate Filament Protein. Accession Number: P47819. Gene ID: 24387. Expression Region: Met1~Met430. Tag Info: N-terminal His Tag. Theoretical MW: 54kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0110