Learn More
Biomatik Corporation Recombinant Human Glucose Transporter 4 (GLUT4)
Supplier:  Biomatik Corporation RPU50709
Encompass_Preferred
Recombinant Human Glucose Transporter 4 (GLUT4) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Glucose Transporter 4 (GLUT4). Target Synonyms: SLC2A4; Solute carrier family 2, facilitated glucose transporter member 4; Glucose transporter type 4, insulin-responsive. Accession Number: P14672. Gene ID: 6517. Expression Region: Arg228~Val292. Tag Info: N-terminal His and GST Tag. Theoretical MW: 41kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0112