Learn More
Biomatik Corporation BSA Conjugated Asymmetrical Dimethylarginine (ADMA)
Supplier:  Biomatik Corporation RPU50856
Encompass_Preferred
BSA Conjugated Asymmetrical Dimethylarginine (ADMA) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: Asymmetrical Dimethylarginine (ADMA). Target Synonyms: N,N-Dimethylarginine; NG,NG-Dimethylarginine dihydrochloride; Asymmetric Dimethylarginine. Tag Info: N-terminal His Tag. Theoretical MW: 202.3Da. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0116