Learn More
Biomatik Corporation Recombinant Human Lipoprotein, a (Lpa)
Supplier:  Biomatik Corporation RPU51144
Encompass_Preferred
Recombinant Human Lipoprotein, a (Lpa) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Lipoprotein, a (Lpa). Target Synonyms: Lp(a); AK38; APOA; LP; Sinking Pre-β-Lipoprotein. Accession Number: P08519. Gene ID: 4018. Expression Region: Asp1719~Arg2038. Tag Info: N-terminal His Tag. Theoretical MW: 39kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0129