Learn More
Biomatik Corporation Recombinant Mouse Diazepam Binding Inhibitor (DBI)
Supplier:  Biomatik Corporation RPU41193
Encompass_Preferred
Recombinant Mouse Diazepam Binding Inhibitor (DBI) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Diazepam Binding Inhibitor (DBI). Target Synonyms: ACBP; ACBD1; CCK-RP; EP; Endozepine; GABA Receptor Modulator; Acyl-Coenzyme A Binding Protein. Accession Number: P31786. Gene ID: 13167. Expression Region: Ser2~Ile87. Tag Info: N-terminal His Tag. Theoretical MW: 13kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9926