Learn More
Biomatik Corporation OVA Conjugated Vitamin D3 (VD3)
Supplier:  Biomatik Corporation RPU51267
Encompass_Preferred
OVA Conjugated Vitamin D3 (VD3) is a purified Conjugated Small Molecules. Purity: >90% by SDS-PAGE. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Universal. Target Name: Vitamin D3 (VD3). Target Synonyms: Cholecalciferol; Toxiferol; Activated 7-Dehydrocholesterol. Tag Info: N-terminal His Tag. Theoretical MW: 384.6Da. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0135