Learn More
Biomatik Corporation Recombinant Human Cathepsin G (CTSG)
Catalog No. 502290151
Encompass_Preferred
Recombinant Human Cathepsin G (CTSG) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cathepsin G (CTSG). Target Synonyms: CG; CTS-G. Accession Number: P08311. Gene ID: 1511. Expression Region: Ile21~Leu255. Tag Info: N-terminal His and GST Tag. Theoretical MW: 57kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0151 Supplier Biomatik Corporation Supplier No. RPU51594
Add to Cart
Add to Cart
Recombinant Human Cathepsin G (CTSG) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cathepsin G (CTSG). Target Synonyms: CG; CTS-G. Accession Number: P08311. Gene ID: 1511. Expression Region: Ile21~Leu255. Tag Info: N-terminal His and GST Tag. Theoretical MW: 57kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.