Learn More
Biomatik Corporation Recombinant Mouse Complement Component 1, Q Subcomponent A (C1qA)
Supplier:  Biomatik Corporation RPU51843
Encompass_Preferred
Recombinant Mouse Complement Component 1, Q Subcomponent A (C1qA) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Complement Component 1, Q Subcomponent A (C1qA). Target Synonyms: Complement Component 1,q Subcomponent,Alpha Polypeptide. Accession Number: P98086. Gene ID: 12259. Expression Region: Glu23~Ala245. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0165