Learn More
Biomatik Corporation Recombinant Mouse Annexin A2 (ANXA2)
Supplier:  Biomatik Corporation RPU52100
Encompass_Preferred
Recombinant Mouse Annexin A2 (ANXA2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Annexin A2 (ANXA2). Target Synonyms: ANX-A2; ANX2L4; CAL1H; LIP2; LPC2; LPC2D; P36; PAP-IV; Protein I; Chromobindin-8; Calpactin-1 heavy chain; Lipocortin II; Placental anticoagulant protein IV. Accession Number: P07356. Gene ID: 12306. Expression Region: Met1~Asp339. Tag Info: N-terminal His Tag. Theoretical MW: 42kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0176