Learn More
Biomatik Corporation Recombinant Human Prostate-specific Membrane Antigen (PMSA)
Supplier:  Biomatik Corporation RPU52153
Encompass_Preferred
Recombinant Human Prostate-specific Membrane Antigen (PMSA) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Prostate-specific Membrane Antigen (PMSA). Target Synonyms: FOLH1; PSM; FGCP; FOLH; GCP2; NAALAD1; GCPII; mGCP; NAALADase I; Glutamate Carboxypeptidase II; Folate Hydrolase 1; Folylpoly-gamma-glutamate carboxypeptidase. Accession Number: Q04609. Gene ID: 2346. Expression Region: Ala274~Leu587. Tag Info: N-terminal His Tag. Theoretical MW: 39kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0178