Learn More
Biomatik Corporation Recombinant Human Kelch Like ECH Associated Protein 1 (KEAP1)
Supplier:  Biomatik Corporation RPU52355
Encompass_Preferred
Recombinant Human Kelch Like ECH Associated Protein 1 (KEAP1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Kelch Like ECH Associated Protein 1 (KEAP1). Target Synonyms: INrf2; KLHL19; Cytosolic inhibitor of Nrf2; Kelch-like protein 19. Accession Number: Q14145. Gene ID: 9817. Expression Region: Leu327~Cys624. Tag Info: N-terminal His Tag. Theoretical MW: 37kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0185