Learn More
Biomatik Corporation Recombinant Human Dipeptidyl Peptidase 6 (DPP6)
Supplier:  Biomatik Corporation RPU52473
Encompass_Preferred
Recombinant Human Dipeptidyl Peptidase 6 (DPP6) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Dipeptidyl Peptidase 6 (DPP6). Target Synonyms: DPPX; Dipeptidylpeptidase 6; Dipeptidyl aminopeptidase-like protein 6; Dipeptidyl aminopeptidase-related protein; Dipeptidyl peptidase IV-like protein. Accession Number: P42658. Gene ID: 1804. Expression Region: Phe667~Asp865. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0193