Learn More
Biomatik Corporation Recombinant Human Isoleucyl tRNA Synthetase (IARS)
Supplier:  Biomatik Corporation RPU52866
Encompass_Preferred
Recombinant Human Isoleucyl tRNA Synthetase (IARS) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Isoleucyl tRNA Synthetase (IARS). Target Synonyms: IARS1; ILRS; OJ; IleRS; IRS; Isoleucine tRNA Ligase 1,Cytoplasmic. Accession Number: P41252. Gene ID: 3376. Expression Region: Met1~Arg500. Tag Info: N-terminal His Tag. Theoretical MW: 62kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0204