Learn More
Biomatik Corporation Recombinant Rat 11-Beta-Hydroxysteroid Dehydrogenase Type 2 (HSD11b2)
Supplier:  Biomatik Corporation RPU52893
Encompass_Preferred
Recombinant Rat 11-Beta-Hydroxysteroid Dehydrogenase Type 2 (HSD11b2) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: 11-Beta-Hydroxysteroid Dehydrogenase Type 2 (HSD11b2). Target Synonyms: AME; AME1; HSD11K; HSD2; SDR9C3; Short Chain Dehydrogenase/Reductase Family 9C,Member 3; NAD-dependent 11-beta-hydroxysteroid dehydrogenase. Accession Number: P50233. Gene ID: 25117. Expression Region: Leu58~Pro393. Tag Info: N-terminal His Tag. Theoretical MW: 41kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0206