Learn More
Biomatik Corporation Recombinant Human Protein Kinase C Theta (PKCq)
Supplier:  Biomatik Corporation RPU52987
Encompass_Preferred
Recombinant Human Protein Kinase C Theta (PKCq) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Protein Kinase C Theta (PKCq). Target Synonyms: PRKC-Q; PRKCQ; PRKCT; NPKC-Theta. Accession Number: Q04759. Gene ID: 5588. Expression Region: Phe380~Phe634. Tag Info: N-terminal His and GST Tag. Theoretical MW: 60kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0212