Learn More
Biomatik Corporation Recombinant Human Farnesoid X Receptor (FXR)
Supplier:  Biomatik Corporation RPU53082
Encompass_Preferred
Recombinant Human Farnesoid X Receptor (FXR) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Farnesoid X Receptor (FXR). Target Synonyms: NR1H4; BAR; FXR; HRR1; RIP14; Nuclear Receptor Subfamily 1,Group H,Member 4; Bile acid receptor; Farnesol receptor HRR-1; Retinoid X receptor-interacting protein 14. Accession Number: Q96RI1. Gene ID: 9971. Expression Region: Leu211~Gln486. Tag Info: Two N-terminal Tags, His-tag and SUMO-tag. Theoretical MW: 53kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0216