Learn More
Biomatik Corporation Recombinant Human ATP Binding Cassette Transporter G2 (ABCG2)
Supplier:  Biomatik Corporation RPU53513
Encompass_Preferred
Recombinant Human ATP Binding Cassette Transporter G2 (ABCG2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: ATP Binding Cassette Transporter G2 (ABCG2). Target Synonyms: CDw338; CD338; ABC-G2; MRX; BCRP1; ABC15; ABCP; BCRP; BMDP; MXR; MXR1; Mitoxantrone resistance-associated protein; Breast Cancer Resistance Protein 1; Urate exporter. Accession Number: Q9UNQ0. Gene ID: 9429. Expression Region: Met1~Pro290. Tag Info: N-terminal His Tag. Theoretical MW: 35kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0226