Learn More
Biomatik Corporation Recombinant Mouse Splicing Factor 3B Subunit 3 (SF3B3)
Supplier:  Biomatik Corporation RPU53793
Encompass_Preferred
Recombinant Mouse Splicing Factor 3B Subunit 3 (SF3B3) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Splicing Factor 3B Subunit 3 (SF3B3). Target Synonyms: RSE1; SAP130; SF3b130; STAF130; Spliceosome-associated protein 130; Pre-mRNA-splicing factor SF3b 130 kDa subunit. Accession Number: Q921M3. Gene ID: 101943. Expression Region: Gly35~Phe210. Tag Info: N-terminal His Tag. Theoretical MW: 23kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0233