Learn More
Biomatik Corporation Recombinant Human Collagen Type VI Alpha 1 (COL6a1)
Supplier:  Biomatik Corporation RPU54375
Encompass_Preferred
Recombinant Human Collagen Type VI Alpha 1 (COL6a1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Collagen Type VI Alpha 1 (COL6a1). Target Synonyms: COL6-A1; OPLL; Collagen Alpha-1(VI)chain. Accession Number: P12109. Gene ID: 1291. Expression Region: Ala828~Ser1022. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0241