Learn More
Biomatik Corporation Recombinant Human Bone Morphogenetic Protein 9 (BMP9)
Supplier:  Biomatik Corporation RPU54653
Encompass_Preferred
Recombinant Human Bone Morphogenetic Protein 9 (BMP9) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Bone Morphogenetic Protein 9 (BMP9). Target Synonyms: GDF2; Growth Differentiation Factor 2. Accession Number: Q9UK05. Gene ID: 2658. Expression Region: Ser32~Gly427. Tag Info: N-terminal His Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0248