Learn More
Biomatik Corporation Human Colony Stimulating Factor 2, Granulocyte Macrophage (GM-CSF), Active Protein
Supplier:  Biomatik Corporation RPU54823
Encompass_Preferred
Human Colony Stimulating Factor 2, Granulocyte Macrophage (GM-CSF), Active Protein is a purified Eukaryotic Protein. Purity: >95% by SDS-PAGE. Host: 293F Cell. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Colony Stimulating Factor 2, Granulocyte Macrophage (GM-CSF). Target Synonyms: CSF2; GMCSF; Sargramostim; Molgramostin; Granulocyte-Macrophage Colony Stimulating Factor. Accession Number: P04141. Gene ID: 1437. Expression Region: Ala18~Glu144. Tag Info: N-terminal His Tag. Theoretical MW: 19-26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0255