Learn More
Biomatik Corporation Human Interleukin 15 (IL15), Active Protein
Supplier:  Biomatik Corporation RPU54856
Encompass_Preferred
Human Interleukin 15 (IL15), Active Protein is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Interleukin 15 (IL15). Accession Number: P40933. Gene ID: 3600. Expression Region: Trp50~Ser162. Tag Info: N-terminal His Tag. Theoretical MW: 14/16/18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0259