Learn More
Biomatik Corporation Recombinant Human Thyroid Peroxidase (TPO)
Supplier:  Biomatik Corporation RPU55230
Encompass_Preferred
Recombinant Human Thyroid Peroxidase (TPO) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Thyroid Peroxidase (TPO). Target Synonyms: TPX; thyroperoxidase. Accession Number: P07202. Gene ID: 7173. Expression Region: Phe19~Arg846. Tag Info: N-terminal His Tag. Theoretical MW: 96kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-0283